Anti-C5a/C5a(desArg)/C5) Mouse Monoclonal Antibody (Biotin) [clone: ]

Lieferant: BIOLEGEND INC
518305 518306
BLEG518305EA 167 EUR
BLEG518305 BLEG518306
Anti-C5a/C5a(desArg)/C5) Mouse Monoclonal Antibody (Biotin) [clone: ]
Antikörper
Type: Primary
Antigen: C5a/C5a(desArg)/C5)
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR