Anti-LAMP1 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: H4A3]

Lieferant: BIOLEGEND INC
328625 328626
BLEG328625EA 294 EUR
BLEG328625 BLEG328626
Anti-LAMP1 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: H4A3]
Antikörper
Type: Primary
Antigen: LAMP1
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, African Green, Baboon, Chimpanzee, Pigtailed Macaque, Rhesus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR