Anti-IL1B Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: ]

Lieferant: BIOLEGEND INC
508207 508208
BLEG508207EA 184 EUR
BLEG508207 BLEG508208
Anti-IL1B Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: ]
Antikörper
Type: Primary
Antigen: IL1B
Clonality: Monoclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR