Anti-CD34 Hamster Monoclonal Antibody (Alexa Fluor® 647) [clone: ]

Lieferant: BIOLEGEND INC
128605 128606
BLEG128605EA 167 EUR
BLEG128605 BLEG128606
Anti-CD34 Hamster Monoclonal Antibody (Alexa Fluor® 647) [clone: ]
Antikörper
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 0
Conjugation: Alexa Fluor® 647
Epitope:
Host: Hamster
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR