Anti-IgM Mouse Monoclonal Antibody (Brilliant Violet® 421)

Lieferant: BIOLEGEND INC
314515 314516
BLEG314515EA 312 EUR
BLEG314515 BLEG314516
Anti-IgM Mouse Monoclonal Antibody (Brilliant Violet® 421)
Antikörper
Type: Primary
Antigen: IgM
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR