Anti-CD4 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: GK1.5]

Lieferant: BIOLEGEND INC
100437 100438
BLEG100437EA 259 EUR
BLEG100437 BLEG100438
Anti-CD4 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: GK1.5]
Antikörper
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 0
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR