Anti-PTPRC Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: HI100]

Lieferant: BIOLEGEND INC
304129 304130
BLEG304129EA 277 EUR
BLEG304129 BLEG304130
Anti-PTPRC Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: HI100]
Antikörper
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Chimpanzee, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR