Anti-CD27 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: O323]

Lieferant: BIOLEGEND INC
302823 302824
BLEG302823EA 303 EUR
BLEG302823 BLEG302824
Anti-CD27 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: O323]
Antikörper
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Baboon, Cynomolgus, Rhesus, Squirrel Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR