Anti-CXCR4 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 12G5]

Lieferant: BIOLEGEND INC
306517 306518
BLEG306517EA 312 EUR
BLEG306517 BLEG306518
Anti-CXCR4 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 12G5]
Antikörper
Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 12G5
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR