Anti-CD86 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: IT2.2]

Lieferant: BIOLEGEND INC
305426 305425
BLEG305426EA 624 EUR
BLEG305426 BLEG305425
Anti-CD86 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: IT2.2]
Antikörper
Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: 0
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR