Anti-CD5 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: UCHT2]

Lieferant: BIOLEGEND INC
300625 300626
BLEG300625EA 288 EUR
BLEG300625 BLEG300626
Anti-CD5 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: UCHT2]
Antikörper
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 0
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR