Anti-CD2 Rat Monoclonal Antibody (Biotin) [clone: ]

Lieferant: BIOLEGEND INC
100103 100104
BLEG100103EA 113 EUR
BLEG100103 BLEG100104
Anti-CD2 Rat Monoclonal Antibody (Biotin) [clone: ]
Antikörper
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR