Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy5®) [clone: ]

Lieferant: BIOLEGEND INC
103109 103110
BLEG103109EA 123 EUR
BLEG103109 BLEG103110
Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy5®) [clone: ]
Antikörper
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone:
Conjugation: PE (Phycoerythrin)-Cy5®
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR