Anti-NK1.1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ]

Lieferant: BIOLEGEND INC
108709 108710
BLEG108709EA 140 EUR
BLEG108709 BLEG108710
Anti-NK1.1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ]
Antikörper
Type: Primary
Antigen: NK-1.1
Clonality: Monoclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR