Anti-ITGA2 Rat Monoclonal Antibody (Biotin) [clone: ]

Lieferant: BIOLEGEND INC
108903 108904
BLEG108903EA 140 EUR
BLEG108903 BLEG108904
Anti-ITGA2 Rat Monoclonal Antibody (Biotin) [clone: ]
Antikörper
Type: Primary
Antigen: ITGA2
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgM kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR