Anti-H-2Kb Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: ]

Lieferant: BIOLEGEND INC
116505 116506
BLEG116505EA 140 EUR
BLEG116505 BLEG116506
Anti-H-2Kb Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: ]
Antikörper
Type: Primary
Antigen: H-2Kb
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR