Anti-CD2 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy5®) [clone: ]

Lieferant: BIOLEGEND INC
300209 300210
BLEG300209EA 193 EUR
BLEG300209 BLEG300210
Anti-CD2 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy5®) [clone: ]
Antikörper
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone:
Conjugation: PE (Phycoerythrin)-Cy5®
Epitope:
Host: Mouse
Isotype:
Reactivity: Baboon, Chimpanzee, Cynomolgus, Human, Pig, Pigtailed Macaque, Pig, Rhesus, Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR