Anti-PTPRC Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: HI30]

Lieferant: BIOLEGEND INC
304005 304006 304038
BLEG304005EA 52 EUR
BLEG304005 BLEG304006 BLEG304038
Anti-PTPRC Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: HI30]
Antikörper
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chimpanzee, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR