Anti-FAS Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: ]

Lieferant: BIOLEGEND INC
305605 305606
BLEG305605EA 174 EUR
BLEG305605 BLEG305606
Anti-FAS Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: ]
Antikörper
Type: Primary
Antigen: CD95 (Fas)
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Baboon, Chimpanzee, Cynomolgus, Human, Pigtailed Macaque, Rhesus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR