Anti-HLA-DR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ]

Lieferant: BIOLEGEND INC
307605 307606
BLEG307605EA 113 EUR
BLEG307605 BLEG307606
Anti-HLA-DR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ]
Antikörper
Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Baboon, Chimpanzee, Cynomolgus, Dog, Human, Marmoset, Rhesus, Tamarin
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR