Anti-IL10 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: ]

Lieferant: BIOLEGEND INC
501403 501404
BLEG501403EA 149 EUR
BLEG501403 BLEG501404
Anti-IL10 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: ]
Antikörper
Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Human, Baboon, Rhesus, Cynomolgus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR