Anti-NGF Mouse Monoclonal Antibody (Unconjugated) [clone: ]

Lieferant: BIOLEGEND INC
509701 509702
BLEG509701EA 153 EUR
BLEG509701 BLEG509702
Anti-NGF Mouse Monoclonal Antibody (Unconjugated) [clone: ]
Antikörper
Type: Primary
Antigen: NGF
Clonality: Monoclonal
Clone:
Conjugation: PURE
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR