Anti-PTPRC Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: HI30]

Lieferant: BIOLEGEND INC
304035 304036
BLEG304035EA 310 EUR
BLEG304035 BLEG304036
Anti-PTPRC Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: HI30]
Antikörper
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR