Anti-HLA-DR Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: L243]

Lieferant: BIOLEGEND INC
307641 307642
BLEG307641EA 344 EUR
BLEG307641 BLEG307642
Anti-HLA-DR Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: L243]
Antikörper
Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 785
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Africangreen, Baboon, Chimpanzee, Common Marmoset, Cotton-Toppedtamarin, Cynomolgus, Pigtailedmacaque, Rhesus, Squirrelmonkey, Dog
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR