Anti-CD34 Mouse Monoclonal Antibody [clone: 561] (Brilliant Violet® 421)

Lieferant: BIOLEGEND INC
343609 343610
BLEG343609EA 320 EUR
BLEG343609 BLEG343610
Anti-CD34 Mouse Monoclonal Antibody [clone: 561] (Brilliant Violet® 421)
Antikörper
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 0
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR