Anti-CD4 Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: RM4-5]

Lieferant: BIOLEGEND INC
100553 100559
BLEG100553EA 269 EUR
BLEG100553 BLEG100559
Anti-CD4 Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: RM4-5]
Antikörper
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 0
Conjugation: Brilliant Violet® 510
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR