Rat IgG2a kappa Isotype Control [clone: RTK2758] (Brilliant Violet® 510)

Lieferant: BIOLEGEND INC
400547 400553
BLEG400547EA 303 EUR
BLEG400547 BLEG400553
Rat IgG2a kappa Isotype Control [clone: RTK2758] (Brilliant Violet® 510)
Antikörper
Type: Control
Antigen: IgG2a κ
Clonality: Monoclonal
Clone: RTK2758
Conjugation: Brilliant Violet® 510
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR