Anti-ITGAX Hamster Monoclonal Antibody (Brilliant Violet® 605) [clone: N418]

Lieferant: BIOLEGEND INC
117333 117334
BLEG117333EA 338 EUR
BLEG117333 BLEG117334
Anti-ITGAX Hamster Monoclonal Antibody (Brilliant Violet® 605) [clone: N418]
Antikörper
Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: N418
Conjugation: Brilliant Violet® 605
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR