Anti-IL7R Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: A019D5]

Lieferant: BIOLEGEND INC
351325 351326
BLEG351325EA 353 EUR
BLEG351325 BLEG351326
Anti-IL7R Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: A019D5]
Antikörper
Type: Primary
Antigen: IL7R
Clonality: Monoclonal
Clone: A019D5
Conjugation: Brilliant Violet® 650
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR