Anti-CCR7 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: G043H7]

Lieferant: BIOLEGEND INC
353223 353224
BLEG353223EA 379 EUR
BLEG353223 BLEG353224
Anti-CCR7 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: G043H7]
Antikörper
Type: Primary
Antigen: CCR7
Clonality: Monoclonal
Clone: G043H7
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Cynomolgus, Rhesus, Baboon
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR