Anti-CD27 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: LG.3A10]

Lieferant: BIOLEGEND INC
124216 124215
BLEG124216EA 471 EUR
BLEG124216 BLEG124215
Anti-CD27 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: LG.3A10]
Antikörper
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: LG.3A10
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Armenian Hamster
Isotype:
Reactivity: Mouse, Rat, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR