Anti-CLEC4C Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 201A]

Lieferant: BIOLEGEND INC
354211 354212
BLEG354211EA 303 EUR
BLEG354211 BLEG354212
Anti-CLEC4C Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 201A]
Antikörper
Type: Primary
Antigen: CLEC4C
Clonality: Monoclonal
Clone: 201A
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR