Anti-HA Epitope Tag Mouse Monoclonal Antibody (Agarose) [clone: 2-2.2.14]

Lieferant: Thermo Fisher Scientific
26181 26182
PIER26181EA 708 EUR
PIER26181 PIER26182
Anti-HA Epitope Tag Mouse Monoclonal Antibody (Agarose) [clone: 2-2.2.14]
Antikörper
Type: Primary
Antigen:
Clonality: Monoclonal
Clone: 2-2.2.14
Conjugation: Agarose
Epitope: HA epitope tag
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR