Anti-IgG1 Goat Polyclonal Antibody (CF™ 405S)

Lieferant: Biotium
20380-250UL 20380-50UL
BTIU20380-250ULEA 385 EUR
BTIU20380-250UL BTIU20380-50UL
Anti-IgG1 Goat Polyclonal Antibody (CF™ 405S)
Antikörper
Goat anti-mouse IgG1 is an affinity-purified antibody that reacts with Fc portion of the heavy chain of mouse IgG1. To minimize cross reactivity, the antibody is cross-adsorbed against other mouse IgG subclasses (IgG2a, IgG2b, IgG3), and human, bovine and rabbit serum proteins. CF™405S is a blue fluorescent dye with an absorption peak wavelength that nearly coincides with the 405 nm blue diode laser line. In addition, the emission peak wavelength of the dye well centers within the blue detection window of BD flow cytometers. As a result, flow cytometry analysis using CF™405S results in much brighter signal compared to Alexa Fluor® 405.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgG1
Clonality: Polyclonal
Clone:
Conjugation: CF405S
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR