Anti-IgG2a Goat Polyclonal Antibody (CF™ 405S)

Lieferant: Biotium
20381-50UL 20381-250UL
BTIU20381-50ULEA 168 EUR
BTIU20381-50UL BTIU20381-250UL
Anti-IgG2a Goat Polyclonal Antibody (CF™ 405S)
Antikörper
Goat anti-mouse IgG2a is an affinity-purified antibody that reacts with Fc portion of the heavy chain of mouse IgG2a. To minimize cross reactivity, the antibody is cross-adsorbed against other mouse IgG subclasses (IgG1, IgG2b, IgG3), and human, bovine and rabbit serum proteins. CF™405S is a blue fluorescent dye with an absorption peak wavelength that nearly coincides with the 405 nm blue diode laser line. In addition, the emission peak wavelength of the dye well centers within the blue detection window of BD flow cytometers. As a result, flow cytometry analysis using CF™405S results in much brighter signal compared to Alexa Fluor® 405.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: CF405S
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR