Anti-MUC16 Mouse Monoclonal Antibody [clone: OC125]

Lieferant: IMMUNOLOGIC
ILM7443-C01 ILM7443-C05 ILM7443-C1 ILM7443-R10 ILM7443-R25
ILM7443-C01EA 233 EUR
ILM7443-C01 ILM7443-C05 ILM7443-C1 VWRKILM7443-R10 VWRKILM7443-R25
Anti-MUC16 Mouse Monoclonal Antibody [clone: OC125]
Antikörper
Anti-CA-125 antibody reacts with epithelioid malignancies of the ovary, papillary serous carcinoma of the cervix, adenocarcinoma of the endometrium, clear cell adenocarcinoma of the bladder, and epithelioid mesothelioma1-4. The antigen is formalin resistant, permitting the detection of ovarian cancer by immunohistochemistry, although serum assays for this protein are widely used to monitor ovarian cancer.5

  • Visualization: cytoplasmic and membranous
  • Control: ovarian serous carcinoma
  • Dilution Range: 1:100-1:500

Refernces:

1. Kabawat S, et al. Int J Gynecol Pathol. 1983; 2:275-285.
2. Davis H, et al. Cancer Res. 1986; 46:6143-6148.
3. Zhou C, et al. Am J Surg Pathol. 1998; 22:113- 20.
4. Mylonas I, et al. Anticancer Res. 2003; 23:1075-80.
5. Fukazawa I, et al. Arch Gynecol Obstet. 1988; 243:41-50.

Type: Primary
Antigen: MUC16
Clonality: Monoclonal
Clone: OC125
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR