Anti-SERPINA1 Rabbit Polyclonal Antibody

Lieferant: IMMUNOLOGIC
ILP2113-C01 ILP2113-C05 ILP2113-C1
ILP2113-C01EA 67.5 EUR
ILP2113-C01 ILP2113-C05 VWRKILP2113-C1
Anti-SERPINA1 Rabbit Polyclonal Antibody
Antikörper
The immunohistochemical staining of Alpha-1-Antitrypsin is considered to be very useful in the study of inherited AAT deficiency, benign and malignant hepatic tumors and yolk sac carcinomas. Positive staining for A-1-Antitrypsin may also be used in detection of benign and malignant lesions of an histiocytic nature. Sensitivity and specificity of the results have made this antibody a useful tool in the screening of patients with cryptogenic cirrhosis or other forms of liver disease with portal fibrosis of uncertain etiology.

  • Visualization: cytoplasmic
  • Control: tonsil
  • Dilution Range: 1:500-1:2000

Type: Primary
Antigen: SERPINA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR