Anti-IgA Rabbit Polyclonal Antibody

Lieferant: IMMUNOLOGIC
ILP2673-C01 ILP2673-C05 ILP2673-C1
ILP2673-C01EA 53.5 EUR
ILP2673-C01 ILP2673-C05 VWRKILP2673-C1
Anti-IgA Rabbit Polyclonal Antibody
Antikörper
Anti-IgA antibody reacts with surface immunoglobulin IgA alpha chains. It is useful when identifying leukemias, plasmacytomas, and B-cell lineage derived Hodgkin’s lymphomas. Due to the restricted expression of heavy and light chains in these diseases, demonstration of B-cell lymphoma/plasmacytoma is aided with this antibody.

  • Visualization: cytoplasmic
  • Control: tonsil
  • Dilution Range: 1:100-1:500

Type: Primary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR