Anti-TSHB Rabbit Polyclonal Antibody

Lieferant: IMMUNOLOGIC
ILP9113-C01 ILP9113-C05 ILP9113-C1
ILP9113-C01EA 154 EUR
ILP9113-C01 ILP9113-C05 VWRKILP9113-C1
Anti-TSHB Rabbit Polyclonal Antibody
Antikörper
Anti-TSH is a useful marker in classification of pituitary tumors and the study of pituitary disease. It reacts with TSH-producing cells (thyrotrophs).

  • Visualization: cytoplasmic
  • Control: pituitary
  • Dilution Range: 1:100-1:500

Type: Primary
Antigen: TSHB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR