Anti-EPCAM Mouse Monoclonal Antibody [clone: VU1D9]

Lieferant: IMMUNOLOGIC
ILM6727-C01 ILM6727-C05 ILM6727-C1
ILM6727-C01EA 219 EUR
ILM6727-C01 ILM6727-C05 ILM6727-C1
Anti-EPCAM Mouse Monoclonal Antibody [clone: VU1D9]
Antikörper
EpCAM is a transmembrane glycoprotein involved in a number of cellular processes including adhesion, proliferation, maintenance of stemness, migration and invasion.3 It is expressed on most, but not all, normal epithelia and their corresponding malignancies. Although its role in tumorigenesis remains to be fully elucidated, it may downregulate immunity and help tumors actively escape from immune surveillance. It has been recognized as a pan-carcinoma antigen marker because of its widespread expression on epithelium and their derived tumors.1 Its antibody is widely used to help distinguishMore Product Information
Bestellen

TECHNISCHE DATEN

Antigen-Symbol EPCAM
Antigen-Name Epithelial cell adhesion molecule
Environmentally Preferable
Antigen-Synonyme CD326|CO-17A|Ly74|TACSTD1|HEA125|MIC18|EGP34|Ep-CAM|MOC31|TACST-1|MH99|17-1A|M4S1|EGP40|KS1/4|KSA|MK-1|TROP1|GA733-2|ESA|EGP-2|323/A3
Antikörper-Typ Primär
Klonalität Monoklonal
Konjugation Unconjugated
Klon VU1D9
Reaktivität Human
Wirt Mouse
Gen-ID 4072
Isotyp IgG1 kappa
Western blot Yes
ELISA Yes
ImmunoChemie Yes
Durchflusszytometrie Yes
Form Concentrated
Molecular weight 40 kDa
Lagertemperatur 2°C - 8°C
Validierte Anwendungen Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR