BrightVision+ Anti-IgG (Poly-HRP)

Lieferant: IMMUNOLOGIC
DPVB110HRP DPVB500HRP DPVB55HRP DPVB999HRP
DPVB110HRPEA 886 EUR
DPVB110HRP DPVB500HRP DPVB55HRP DPVB999HRP
BrightVision+ Anti-IgG (Poly-HRP)
Antikörper
BrightVision + histostaining reagents utilise a novel controlled polymerisation technology to prepare polymeric HRP linker antibody conjugates. Comparing to conventional biotin-streptavidin based detection kits, these kits have the advantages of higher amplification power, biotin free and more consistent immunostaining outcomes on archival tissues and on difficult-to-work antibodies.

  • High-quality secondary antibody for IHC applications, HRP labelled.
  • Antigen specificity: Anti-Mouse IgG (H+L), Anti-Rabbit IgG (H+L)
  • Kit contains post-antibody blocking (ready-to-use) and poly-HRP-anti-mouse/rabbit IgG (ready-to-use)

Colour version available (see VWRKS/C-DPVB-HRP and similar articles)

Biotin-free
Ready-to-use

Standards: Meets CE/IVD standards.

Bestellinformation: Packages above 110 ml contain small empty bottles so users can create smaller working volumes and prevent possible contamination of the whole lot.

Lieferumfang: Shipped with blue ice. Store between 2 to 8 °C.

Bitte beachten Irritating to eyes, respiratory systems and skin.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR