Anti-MME Mouse Monoclonal Antibody (CF405S) [clone: CB-CALLA]

Lieferant: Biotium
BNC040146-500 BNC040146-100
BNC040146-500EA 560 EUR
BNC040146-500 BNC040146-100
Anti-MME Mouse Monoclonal Antibody (CF405S) [clone: CB-CALLA]
Antikörper
Recognizes a 100 kDa glycoprotein, identified as CD10, also known as Common Acute Lymphocytic Leukemia Antigen (CALLA). It is a cell surface enzyme with neutral metalloendopeptidase activity, which inactivates a variety of biologically active peptides. CD10 is expressed on the cells of lymphoblastic, Burkitt's, and follicular germinal center lymphomas, and on cells from patients with chronic myelocytic leukemia (CML). It is also expressed on the surface of normal early lymphoid progenitor cells, immature B cells within adult bone marrow and germinal center B cells within lymphoid tissue. CD10 is also present on breast myoepithelial cells, bile canaliculi, fibroblasts, with especially high expression on the brush border of kidney and gut epithelial cells.

Type: Primary
Antigen: MME
Clonality: Monoclonal
Clone: CB-CALLA
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR