Anti-UCHL1 Mouse Monoclonal Antibody (Biotin) [clone: 13C4]

Lieferant: Biotium
BNCB0147-500 BNCB0147-100
BNCB0147-500EA 560 EUR
BNCB0147-500 BNCB0147-100
Anti-UCHL1 Mouse Monoclonal Antibody (Biotin) [clone: 13C4]
Antikörper
This MAb reacts with a protein of 20-30 kDa, identified as PGP9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UchL1). Initially, PGP9.5 expression in normal tissues was reported in neurons and neuroendocrine cells but later it was found in distal renal tubular epithelium, spermatogonia, Leydig cells, oocytes, melanocytes, prostatic secretory epithelium, ejaculatory duct cells, epididymis, mammary epithelial cells, Merkel cells, and dermal fibroblasts. Furthermore, immunostaining for PGP9.5 has been shown in a wide variety of mesenchymal neoplasms as well. A mutation in PGP9.5 gene is believed to cause a form of Parkinson's disease.

Type: Primary
Antigen: UCHL1
Clonality: Monoclonal
Clone: 13C4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR