Anti-CKB Mouse Monoclonal Antibody (CF594) [clone: 2ba6]

Lieferant: Biotium
BNC940196-500 BNC940196-100
BNC940196-500EA 560 EUR
BNC940196-500 BNC940196-100
Anti-CKB Mouse Monoclonal Antibody (CF594) [clone: 2ba6]
Antikörper
Creatine kinases (CK) are a large family of isoenzymes that regulate levels of ATP in subcellular compartments, where they provide ATP at sites of fluctuating energy demand by the transfer of phosphates between creatine and adenine nucleotides. CKs provide the energy of phosphate hydrolysis necessary to drive the normal function of many cellular systems. In cells, the cytosolic CK enzymes consist of two subunits, which can be either B (brain type) or M (muscle type). There are three different isoenzymes: CKMM, CKBB and CKMB. This MAb recognizes the CKBB isoenzyme and does not react with theMore Product Information
Bestellen

TECHNISCHE DATEN

Antigen-Symbol CKB CKB
Environmentally Preferable
Antigen-Name Creatine Phosphokinase Creatine Phosphokinase
Antikörper-Typ Primär Primär
Klonalität Monoklonal Monoklonal
Konjugation CF® 594 CF® 594
Klon Clone 2ba6 Clone 2ba6
Reaktivität Human Human
Wirt Mouse Mouse
Gen-ID 173724 173724
Isotyp IgG1 kappa IgG1 kappa
ImmunoChemie Yes Yes
ImmunFluoreszenz Yes
Konzentration 0,1 mg/ml 0,1 mg/ml
Lagerungspuffer PBS/0,1% BSA/0,05% azide PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR