Anti-ERBB2 Mouse Monoclonal Antibody (CF488A) [clone: HRB2/258]

Lieferant: Biotium
BNC880258-500 BNC880258-100
BNC880258-500EA 560 EUR
BNC880258-500 BNC880258-100
Anti-ERBB2 Mouse Monoclonal Antibody (CF488A) [clone: HRB2/258]
Antikörper
Recognizes a protein of 185 kDa, which is identified as c-erbB-2/HER-2/neu. Its epitope is localized in the extracellular domain. C-erbB-2/HER-2 is a member of the EGFR family. This MAb is specific and shows minimal cross-reaction with other members of the EGFR-family. Receptors of this family are located on the plasma membrane and consist of an extracellular ligand-binding domain that is connected to a large intracellular domain by a single transmembrane sequence. C-erbB-2/HER-2 protein is over-expressed in a variety of carcinomas especially those of breast and ovary.

Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: HRB2/258
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR