Anti-ESR1 Mouse Monoclonal Antibody (CF568) [clone: AER314]

Lieferant: Biotium
BNC680406-500 BNC680406-100
BNC680406-500EA 560 EUR
BNC680406-500 BNC680406-100
Anti-ESR1 Mouse Monoclonal Antibody (CF568) [clone: AER314]
Antikörper
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. Epitope of this MAb is mapped between aa120-170. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: AER314
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cow, Human, Mouse, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR