Anti-PGR Mouse Monoclonal Antibody (CF405S) [clone: PR484]

Lieferant: Biotium
BNC040484-500 BNC040484-100
BNC040484-500EA 560 EUR
BNC040484-500 BNC040484-100
Anti-PGR Mouse Monoclonal Antibody (CF405S) [clone: PR484]
Antikörper
This MAb is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81 kDa) and PR-B (116 kDa). Expression of PgR has been suggested to reflect a intact estrogen regulatory machinery and therefore, predict better clinical response to endocrine therapy than ER alone. It is excellent for immunohistochemical staining of formalin/paraffin tissues.

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: PR484
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

TECHNISCHE DATEN

Antigen-Symbol PGR
Environmentally Preferable
Antigen-Name Progesterone Receptor
Antigen-Synonyme PR|PRA|Progesterone receptor form A|Nuclear receptor subfamily 3 group C member 3|NR3C3|PRB|PGR|Progesterone receptor form B
Antikörper-Typ Primär
Klonalität Monoklonal
Konjugation CF® 405S
Klon Clone PR484
Reaktivität Human
Wirt Mouse
Gen-ID 2905
Isotyp IgG1 kappa
ImmunoChemie Yes
ImmunFluoreszenz Yes
Konzentration 0,1 mg/ml
Lagerungspuffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR