Anti-MSN Mouse Monoclonal Antibody (CF488A) [clone: MSN492]
Lieferant: Biotium
BNC880492-500 BNC880492-100
BNC880492-500EA 560 EUR
BNC880492-500 BNC880492-100
Anti-MSN Mouse Monoclonal Antibody (CF488A) [clone: MSN492]
Antikörper
Recognizes 78 kDa moesin protein. Moesin, a member of the talin-4.1 superfamily, is a linking protein of the sub-membranous actin cytoskeleton. It is expressed in variable amounts in cells of different phenotypes such as macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. The ERM proteins, ezrin, radixin, and moesin are involved in a variety of cellular functions, such as cell adhesion, migration, and the organization of cell surface structures, and are highly homologous, both in protein sequence and in functional activity, with merlin/schwannomin, a neurofibromatosis-2-associated tumor-suppressor protein. Cell lines of epithelial and mesothelial origin contain both moesin and radixin whereas cells of endothelial and lymphoid origin express moesin.
Type: Primary
Antigen: MSN
Clonality: Monoclonal
Clone: MSN492
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human