Anti-ANPEP Mouse Monoclonal Antibody (CF568) [clone: APN/514]

Lieferant: Biotium
BNC680514-500 BNC680514-100
BNC680514-500EA 560 EUR
BNC680514-500 BNC680514-100
Anti-ANPEP Mouse Monoclonal Antibody (CF568) [clone: APN/514]
Antikörper
This MAb recognizes an extracellular epitope of an integral membrane glycoprotein of 150 kDa, identified as CD13. This antigen is present on most cells of myeloid origin including granulocytes, monocytes, mast cells, and GM-progenitor cells. It is also expressed by the majority of AML, CML in myeloid blast crisis, and in a smaller fraction of lymphoid leukemias. It is absent from normal lymphocytes, platelets and erythrocytes. CD13 is also present on fibroblasts; endothelial cells, epithelial cells from renal proximal tubules and intestinal brush border, bone marrow stromal cells, osteoclasts, and cells lining bile duct canaliculi. CD13 is identical to aminopeptidase N (APN), a prominent membrane-bound metalloprotease present on the surface of intestinal brush border and renal tubules. CD13 plays a role in metabolism of biologically active peptides, in phagocytosis, and in bactericidal/tumoricidal activities. It also serves as a receptor for human coronaviruses (HCV). The lineage-restricted pattern of expression of CD13 within the hemopoietic compartment suggests that it may be important in myeloid cell differentiation.

Type: Primary
Antigen: ANPEP
Clonality: Monoclonal
Clone: APN/514
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR