Anti-AFP Mouse Monoclonal Antibody (Biotin) [clone: C2]

Lieferant: Biotium
BNCB0523-500 BNCB0523-100
BNCB0523-500EA 560 EUR
BNCB0523-500 BNCB0523-100
Anti-AFP Mouse Monoclonal Antibody (Biotin) [clone: C2]
Antikörper
This antibody recognizes an oncofetal glycoprotein with a single chain of 70 kDa, which is identified as alpha fetoprotein (AFP) (ISOBM TD-2 workshop). This MAb is highly specific to AFP and shows no cross-reaction with other oncofetal antigens or serum albumin. AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.

Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: C2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR